Assay Details
| Gene Symbol: VKORC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124445.4 | XP_003124493.1 | 433 | 84 | ||
| GenBank mRNA | AK396762.1 | - | 2 - 3 | 432 | 84 | |
More Information
Set Membership: |
|

VKORC1L1
Genomic Map