Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356063.4 | XP_003356111.1 | 645 | 86 | ||
| XM_005655967.3 | XP_005656024.1 | 613 | 86 | |||
| XM_005655968.3 | XP_005656025.1 | 645 | 86 | |||
| GenBank mRNA | AK396870.1 | - | 4 - 5 | 590 | 86 | |
| AK400138.1 | - | 4 - 5 | 590 | 86 | ||
More Information
Set Membership: |
|

SYT5
Genomic Map