Assay Details
| Gene Symbol: SCUBE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977806.2 | XP_013833260.2 | 3561 | 71 | ||
| XM_021098614.1 | XP_020954273.1 | 3513 | 71 | |||
| XM_021098615.1 | XP_020954274.1 | 3399 | 71 | |||
| GenBank mRNA | AK396947.1 | - | 7 - 8 | 1022 | 71 | |
| KP784762.1 | - | 20 - 21 | 3179 | 71 | ||
More Information
Set Membership: |
|

SCUBE3
Genomic Map