Assay Details
Gene Symbol: FAM117A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003358095.5 | XP_003358143.2 | 416 | 69 | ||
GenBank mRNA | AK397026.1 | - | 1 - 2 | 139 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
