Assay Details
| Gene Symbol: SDR9C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355478.4 | XP_003355526.4 | 8261 | 78 | ||
| XM_021091808.1 | XP_020947467.1 | 7127 | 78 | |||
| GenBank mRNA | AK397372.1 | - | 3 - 4 | 644 | 78 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Sus scrofa  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 SDR9C7
SDR9C7 Genomic Map
Genomic Map Back To Top
 Back To Top