Assay Details
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482681.4 | XP_003482729.1 | 1193 | 71 | ||
XM_005656682.3 | XP_005656739.1 | 806 | 71 | |||
XM_013989572.2 | XP_013845026.1 | 940 | 71 | |||
XM_013989573.2 | XP_013845027.1 | 940 | 71 | |||
GenBank mRNA | AK397583.1 | - | 2 - 3 | 525 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
