Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927669.7 | XP_001927704.4 | 1541 | 83 | ||
GenBank mRNA | AK397615.1 | - | 6 - 7 | 476 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
