Assay Details
Gene Symbol: KCNJ15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005670338.3 | XP_005670395.1 | 2161 | 139 | ||
XM_005670339.3 | XP_005670396.1 | 2156 | 139 | |||
XM_005670340.3 | XP_005670397.1 | 2760 | 139 | |||
XM_005670341.3 | XP_005670398.1 | 3942 | 139 | |||
XM_013982588.2 | XP_013838042.1 | 1725 | 139 | |||
XM_021070979.1 | XP_020926638.1 | 2048 | 139 | |||
XM_021070981.1 | XP_020926640.1 | 1957 | 139 | |||
XM_021070982.1 | XP_020926641.1 | 1843 | 139 | |||
XM_021070983.1 | XP_020926642.1 | 2034 | 139 | |||
XM_021070984.1 | XP_020926643.1 | 1734 | 139 | |||
GenBank mRNA | AK398047.1 | - | 1 | 226 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
