Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021063211.1 | XP_020918870.1 | 870 | 67 | ||
XM_021063212.1 | XP_020918871.1 | 860 | 67 | |||
GenBank mRNA | AK398198.1 | - | 7 - 8 | 758 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
