Assay Details
| Gene Symbol: IL20RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992536.2 | XP_013847990.2 | 1324 | 104 | ||
| XM_021087494.1 | XP_020943153.1 | 3278 | 104 | |||
| XM_021087511.1 | XP_020943170.1 | 2978 | 104 | |||
| GenBank mRNA | AK398612.1 | - | 1 | 224 | 104 | |
More Information
Set Membership: |
|

IL20RA
Genomic Map