Assay Details
| Gene Symbol: MARCHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653890.3 | XP_005653947.1 | 2423 | 83 | ||
| GenBank mRNA | AK398630.1 | - | 2 - 3 | 341 | 83 | |
More Information
Set Membership: |
|

MARCHF10
Genomic Map