Assay Details
Gene Symbol: PIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673107.3 | XP_005673164.2 | 266 | 110 | ||
GenBank mRNA | AK399222.1 | - | 2 - 3 | 247 | 110 | |
AK399235.1 | - | 2 - 3 | 249 | 110 | ||
AK399255.1 | - | 2 - 3 | 246 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
