Assay Details
| Gene Symbol: C15H2orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133545.5 | XP_003133593.2 | 279 | 114 | ||
| GenBank mRNA | AK400090.1 | - | 1 - 2 | 179 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003133545.5 transcripts.
Set Membership: |
|

C15H2orf88
Genomic Map