Assay Details
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124905.5 | XP_003124953.2 | 658 | 102 | ||
XM_021087196.1 | XP_020942855.1 | 573 | 102 | |||
GenBank mRNA | AY609631.1 | - | 5 - 6 | 605 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
