Assay Details
| Gene Symbol: REP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126410.4 | XP_003126458.1 | 1944 | 117 | ||
| GenBank mRNA | AY610002.1 | - | 1 | 815 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: REP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126410.4 | XP_003126458.1 | 1944 | 117 | ||
| GenBank mRNA | AY610002.1 | - | 1 | 815 | 117 | |
Set Membership: |
|