Assay Details
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005654708.3 | XP_005654765.1 | 1332 | 74 | ||
XM_021084115.1 | XP_020939774.1 | 1039 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
