Assay Details
| Gene Symbol: BNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128450.4 | XP_003128498.2 | 2321 | 101 | ||
| XM_021098879.1 | XP_020954538.1 | 2873 | 101 | |||
More Information
Set Membership: |
|

BNC1
Genomic Map