Assay Details
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482086.4 | XP_003482134.1 | 658 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482086.4 | XP_003482134.1 | 658 | 85 |