Assay Details
Gene Symbol: LRFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481830.4 | XP_003481878.2 | 1073 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003481830.4 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
