Assay Details
Gene Symbol: LOC100620470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355459.4 | XP_003355507.1 | 870 | 63 | ||
XM_005663895.3 | XP_005663952.1 | 726 | 63 | |||
GenBank mRNA | AK232303.1 | - | 3 - 4 | 689 | 63 | |
AK233183.1 | - | 3 - 4 | 696 | 63 | ||
AK233628.1 | - | 4 - 5 | 953 | 63 | ||
AK394106.1 | - | 3 - 4 | 725 | 63 | ||
AK394142.1 | - | 3 - 4 | 725 | 63 | ||
AK398963.1 | - | 3 - 4 | 802 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
