Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124561.6 | XP_003124609.3 | 738 | 62 | ||
| XM_021086520.1 | XP_020942179.1 | 728 | 62 | |||
| XM_021086521.1 | XP_020942180.1 | 737 | 62 | |||
| GenBank mRNA | AK232522.1 | - | 4 - 5 | 686 | 62 | |
| AK393717.1 | - | 4 - 5 | 721 | 62 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map