Assay Details
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123321.6 | XP_003123369.1 | 589 | 80 | ||
XM_005652853.3 | XP_005652910.1 | 564 | 80 | |||
GenBank mRNA | AK232558.1 | - | 3 - 4 | 392 | 80 | |
AK350433.1 | - | 3 - 4 | 378 | 80 | ||
AK350775.1 | - | 3 - 4 | 367 | 80 | ||
AK397370.1 | - | 3 - 4 | 528 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
