Assay Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099112.1 | XP_020954771.1 | 3467 | 67 | ||
| XM_021099120.1 | XP_020954779.1 | 2255 | 67 | |||
| XM_021099126.1 | XP_020954785.1 | 2362 | 67 | |||
| GenBank mRNA | AK232608.1 | - | 1 | 718 | 67 | |
More Information
Set Membership: |
|

TSHZ1
Genomic Map