Assay Details
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127468.5 | XP_003127516.2 | 851 | 61 | ||
XM_005655974.3 | XP_005656031.1 | 765 | 61 | |||
GenBank mRNA | AK232617.1 | - | 1 - 2 | 174 | 61 | |
AK351286.1 | - | 1 - 2 | 174 | 61 | ||
AK396222.1 | - | 3 - 4 | 578 | 61 | ||
AY609827.1 | - | 3 - 4 | 538 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
