Assay Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003354653.5 | XP_003354701.1 | 295 | 132 | ||
GenBank mRNA | AK232633.1 | - | 2 - 3 | 257 | 132 | |
AK348375.1 | - | 2 - 3 | 259 | 132 | ||
AK396001.1 | - | 3 - 4 | 435 | 132 | ||
AK399050.1 | - | 2 - 3 | 273 | 132 | ||
AK400676.1 | - | 2 - 3 | 259 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
