Assay Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003354653.5 | XP_003354701.1 | 416 | 60 | ||
GenBank mRNA | AK232633.1 | - | 3 - 4 | 378 | 60 | |
AK234196.1 | - | 4 - 5 | 513 | 60 | ||
AK348375.1 | - | 3 - 4 | 380 | 60 | ||
AK395878.1 | - | 4 - 5 | 515 | 60 | ||
AK396001.1 | - | 4 - 5 | 556 | 60 | ||
AK399050.1 | - | 3 - 4 | 394 | 60 | ||
AK400676.1 | - | 3 - 4 | 380 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
