Assay Details
Gene Symbol: PAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355889.4 | XP_003355937.1 | 1453 | 58 | ||
XM_005664597.3 | XP_005664654.1 | 1569 | 58 | |||
XM_005664598.3 | XP_005664655.1 | 3407 | 58 | |||
XM_013998501.2 | XP_013853955.1 | 1815 | 58 | |||
XM_021094361.1 | XP_020950020.1 | 1598 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
