Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021063404.1 | XP_020919063.1 | 1087 | 73 | ||
XM_021063405.1 | XP_020919064.1 | 1091 | 73 | |||
XM_021063407.1 | XP_020919066.1 | 684 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
