Assay Details
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087264.1 | XP_020942923.1 | 1857 | 59 | ||
XM_021087266.1 | XP_020942925.1 | 1855 | 59 | |||
XM_021087267.1 | XP_020942926.1 | 1856 | 59 | |||
XM_021087268.1 | XP_020942927.1 | 1856 | 59 | |||
XM_021087269.1 | XP_020942928.1 | 1853 | 59 | |||
XM_021087270.1 | XP_020942929.1 | 1854 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
