Assay Details
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925695.7 | XP_001925730.4 | 1678 | 117 | ||
XM_005657056.3 | XP_005657113.2 | 1858 | 117 | |||
XM_005657060.3 | XP_005657117.2 | 1699 | 117 | |||
XM_013990076.2 | XP_013845530.2 | 1859 | 117 | |||
XM_021071324.1 | XP_020926983.1 | 1519 | 117 | |||
XM_021071325.1 | XP_020926984.1 | 1696 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
