Assay Details
Gene Symbol: PIGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003353561.4 | XP_003353609.1 | 1168 | 67 | ||
XM_005654517.3 | XP_005654574.1 | 943 | 67 | |||
XM_005654518.3 | XP_005654575.1 | 1579 | 67 | |||
XM_021065420.1 | XP_020921079.1 | 1601 | 67 | |||
XM_021065447.1 | XP_020921106.1 | 1173 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
