Assay Details
Gene Symbol: AIFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005657396.3 | XP_005657453.2 | 836 | 66 | ||
XM_005657397.3 | XP_005657454.2 | 836 | 66 | |||
XM_005657398.3 | XP_005657455.2 | 833 | 66 | |||
XM_005657399.3 | XP_005657456.2 | 746 | 66 | |||
XM_005657400.3 | XP_005657457.2 | 743 | 66 | |||
XM_013990430.2 | XP_013845884.2 | 747 | 66 | |||
XM_013990431.2 | XP_013845885.2 | 628 | 66 | |||
XM_021073860.1 | XP_020929519.1 | 746 | 66 | |||
XM_021073861.1 | XP_020929520.1 | 743 | 66 | |||
XM_021073863.1 | XP_020929522.1 | 628 | 66 | |||
XM_021073864.1 | XP_020929523.1 | 653 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
