Assay Details
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003132121.4 | XP_003132169.2 | 1209 | 93 | ||
XM_013981478.2 | XP_013836932.2 | 1048 | 93 | |||
XM_013981480.2 | XP_013836934.2 | 1069 | 93 | |||
XM_021071680.1 | XP_020927339.1 | 1368 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
