Assay Details
Gene Symbol: LOC100738909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088366.1 | XP_020944025.1 | 570 | 110 | ||
XM_021088367.1 | XP_020944026.1 | 1541 | 110 | |||
XM_021088368.1 | XP_020944027.1 | 895 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
