Assay Details
Gene Symbol: LOC100738909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088366.1 | XP_020944025.1 | 735 | 112 | ||
XM_021088367.1 | XP_020944026.1 | 1706 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
