Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130510.5 | XP_003130558.1 | 1809 | 72 | ||
| XM_013980028.2 | XP_013835482.1 | 1677 | 72 | |||
More Information
Set Membership: |
|

DUSP10
Genomic Map