Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927321.6 | XP_001927356.3 | 278 | 71 | ||
| XM_003355032.4 | XP_003355080.1 | 170 | 71 | |||
| XM_021089190.1 | XP_020944849.1 | 71 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001927321.6, XM_003355032.4, XM_021089190.1 transcripts.
Set Membership: |
|

ELOC
Genomic Map