Assay Details
| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078225.1 | XP_020933884.1 | 1249 | 60 | ||
| XM_021078226.1 | XP_020933885.1 | 991 | 60 | |||
More Information
Set Membership: |
|

SLCO4A1
Genomic Map