Assay Details
Gene Symbol: FUT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081229.1 | XP_020936888.1 | 2440 | 87 | ||
XM_021081230.1 | XP_020936889.1 | 2236 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021081229.1, XM_021081230.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
