Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013999004.2 | XP_013854458.2 | 1167 | 90 | ||
XM_013999005.2 | XP_013854459.2 | 1132 | 90 | |||
XM_013999006.2 | XP_013854460.2 | 710 | 90 | |||
XM_013999007.2 | XP_013854461.2 | 612 | 90 | |||
XM_013999008.2 | XP_013854462.2 | 575 | 90 | |||
XM_013999009.2 | XP_013854463.2 | 674 | 90 | |||
XM_013999010.2 | XP_013854464.2 | 1115 | 90 | |||
XM_021095417.1 | XP_020951076.1 | 597 | 90 | |||
XM_021095418.1 | XP_020951077.1 | 659 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
