Assay Details
Gene Symbol: P2RX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021074039.1 | XP_020929698.1 | 1069 | 84 | ||
XM_021074040.1 | XP_020929699.1 | 1069 | 84 | |||
XM_021074041.1 | XP_020929700.1 | 897 | 84 | |||
XM_021074042.1 | XP_020929701.1 | 755 | 84 | |||
XM_021074043.1 | XP_020929702.1 | 784 | 84 | |||
XM_021074044.1 | XP_020929703.1 | 798 | 84 | |||
XM_021074045.1 | XP_020929704.1 | 604 | 84 | |||
XM_021074046.1 | XP_020929705.1 | 838 | 84 | |||
XM_021074047.1 | XP_020929706.1 | 798 | 84 | |||
XM_021074048.1 | XP_020929707.1 | 554 | 84 | |||
XM_021074049.1 | XP_020929708.1 | 455 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
