Assay Details
| Gene Symbol: B3GNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988607.2 | XP_013844061.1 | 1259 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013988607.2 transcripts.
Set Membership: |
|

B3GNT8
Genomic Map