Assay Details
Gene Symbol: KLHL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666300.3 | XP_005666357.2 | 187 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005666300.3 transcripts.
Set Membership: |
![]() ![]() |
