Assay Details
| Gene Symbol: RHOG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659070.3 | XP_005659127.1 | 120 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005659070.3 transcripts.
Set Membership: |
|

RHOG2
Genomic Map