Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021070144.1 | XP_020925803.1 | 119 | 89 | ||
XM_021070151.1 | XP_020925810.1 | 119 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021070144.1, XM_021070151.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
