Assay Details
| Gene Symbol: TRMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091373.1 | XP_020947032.1 | 544 | 61 | ||
| XM_021091374.1 | XP_020947033.1 | 756 | 61 | |||
| XM_021091375.1 | XP_020947034.1 | 545 | 61 | |||
More Information
Set Membership: |
|

TRMU
Genomic Map