Assay Details
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003354775.4 | XP_003354823.1 | 1613 | 78 | ||
XM_005662467.3 | XP_005662524.2 | 1618 | 78 | |||
XM_013996029.2 | XP_013851483.2 | 1619 | 78 | |||
XM_021087380.1 | XP_020943039.1 | 905 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
