Assay Details
Gene Symbol: LOC100523684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013979484.2 | XP_013834938.1 | 2557 | 109 | ||
XM_021062993.1 | XP_020918652.1 | 1036 | 109 | |||
XM_021062994.1 | XP_020918653.1 | 960 | 109 | |||
XM_021062995.1 | XP_020918654.1 | 1167 | 109 | |||
XM_021062996.1 | XP_020918655.1 | 1090 | 109 | |||
XM_021062997.1 | XP_020918656.1 | 1014 | 109 | |||
XM_021062998.1 | XP_020918657.1 | 841 | 109 | |||
XM_021062999.1 | XP_020918658.1 | 2387 | 109 | |||
XM_021063000.1 | XP_020918659.1 | 953 | 109 | |||
XM_021063001.1 | XP_020918660.1 | 2441 | 109 | |||
XM_021063002.1 | XP_020918661.1 | 813 | 109 | |||
XM_021063004.1 | XP_020918663.1 | 384 | 109 | |||
XM_021063005.1 | XP_020918664.1 | 502 | 109 | |||
XM_021063006.1 | XP_020918665.1 | 392 | 109 | |||
XM_021063007.1 | XP_020918666.1 | 1015 | 109 | |||
XM_021063008.1 | XP_020918667.1 | 939 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
