Assay Details
Gene Symbol: LOC100523684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013979484.2 | XP_013834938.1 | 2300 | 91 | ||
XM_021062993.1 | XP_020918652.1 | 779 | 91 | |||
XM_021062994.1 | XP_020918653.1 | 703 | 91 | |||
XM_021062995.1 | XP_020918654.1 | 910 | 91 | |||
XM_021062996.1 | XP_020918655.1 | 833 | 91 | |||
XM_021062997.1 | XP_020918656.1 | 757 | 91 | |||
XM_021062998.1 | XP_020918657.1 | 584 | 91 | |||
XM_021062999.1 | XP_020918658.1 | 2130 | 91 | |||
XM_021063000.1 | XP_020918659.1 | 696 | 91 | |||
XM_021063001.1 | XP_020918660.1 | 2184 | 91 | |||
XM_021063002.1 | XP_020918661.1 | 556 | 91 | |||
XM_021063005.1 | XP_020918664.1 | 245 | 91 | |||
XM_021063007.1 | XP_020918666.1 | 758 | 91 | |||
XM_021063008.1 | XP_020918667.1 | 682 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021063005.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
