Assay Details
Gene Symbol: LOC100523684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062993.1 | XP_020918652.1 | 606 | 129 | ||
XM_021062994.1 | XP_020918653.1 | 530 | 129 | |||
XM_021062995.1 | XP_020918654.1 | 737 | 129 | |||
XM_021062996.1 | XP_020918655.1 | 660 | 129 | |||
XM_021062997.1 | XP_020918656.1 | 584 | 129 | |||
XM_021062998.1 | XP_020918657.1 | 411 | 129 | |||
XM_021062999.1 | XP_020918658.1 | 1957 | 129 | |||
XM_021063000.1 | XP_020918659.1 | 523 | 129 | |||
XM_021063001.1 | XP_020918660.1 | 2011 | 129 | |||
XM_021063002.1 | XP_020918661.1 | 383 | 129 | |||
XM_021063007.1 | XP_020918666.1 | 585 | 129 | |||
XM_021063008.1 | XP_020918667.1 | 509 | 129 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
